Coventry™ Sealed Polyester Swabs
Actual product may vary
Chemtronics
Coventry™ Sealed Polyester Swabs
Exceptionally clean swab that won't fall apart
Have Questions?
Catalog NumberLength (Metric)
FIS19-003-475147 mm
Catalog number FIS19-003-475
Price (SGD)
54.00
Pack
Length (Metric):
147 mm
Price (SGD)
54.00
Pack
Feature a head that's thermoformed and welded to the handle in a single step using a proprietary process.
  • Polypropylene handle is 3.1mm in width (.12 in.)
  • Knit polyester head measures 18.9 × 5.4mm (.75 × 21 in.)
  • Swab: 14.7cm (5.8 in.)
  • Acid and solvent resistant, including acetone
  • Excellent strength
  • High absorbency and absorbency rate
  • Excellent solvent compatibility
  • Do not generate loose fibers or particles
  • No adhesives or binders are used in thecontruction
  • Good for tight tolerance work
  • Micro-fiber has increased number of microspaces to trap more particles, regardless of wet or dry
  • Available Aqua-Prime™ processed
Can be used to:
  • Validate Biogenetic/Pharmaceutical Surfaces
  • Clean Disk Drive Components
  • Clean Sensitive Surfaces in Optics
  • Clean Aerospace Hardware
Specifications
Certifications/ComplianceISO, Class 4, 10
ConstructionSealed
Diameter (English) Handle0.12 in.
Length (English)5.8 in.
For Use With (Application)Can be used to validate biogenetic/pharmaceutical surfaces, clean disk drive components, clean sensitive surfaces in optics, clean aerospace hardware
Product TypeSwab
Diameter (Metric) Handle3 mm
Length (Metric)147 mm
MaterialPolypropylene handle, Knit polyester head
Thickness (Metric)4.8 mm
Length (English) Tip0.75 in.
Length (Metric) Tip18.9 mm
Width (Metric) Tip20 cm
Width (English) Tip0.146 in.
Handle MaterialPolypropylene
Tip MaterialKnit Polyester, Aqua Prime™ processed
Unit SizePack