Durx™ 670 Cleanroom Wipers
Actual product may vary
Berkshire
Durx™ 670 Cleanroom Wipers
For ISO Class 5 to 8 (100 to 100000) Cleanroom Class applications
Have Questions?
Catalog NumberDimensions (L x W)Packaging Quantity
FIS17-987-79A7 x 7 in. (17.78 x 17.78 cm)300/Pk.
Catalog number FIS17-987-79A
Price (SGD)
58.00
Pack
Dimensions (L x W):
7 x 7 in. (17.78 x 17.78 cm)
Packaging Quantity:
300/Pk.
Price (SGD)
58.00
Pack
  • Made from mechanically hydroentangled 55% cellulose/45% polyester; no secondary binders
  • Steam autoclavable for use in aseptic areas
  • Work well with IPA and other solvents
  • Smooth, highly sorbent and durable; general-purpose wiper
  • Low tendency to shed particles; low extractables
  • Also available as wafer separators for horizontal wafer stacking
Specifications
Product TypeCleanroom Wipe
Length (English)7 in.
Length (Metric)17.78 cm
Width (English)7 in.
Width (Metric)17.78 cm
MaterialPolyester Cellulose
CompatibilityCommon cleaning and disinfecting solutions
AutoclavableAutoclavable
Basis Weight67.6
Certifications/ComplianceRoHS 2002/05, REACH, USP
ColorWhite
ConstructionNonwoven
Dimensions (L x W)7 x 7 in. (17.78 x 17.78 cm)
For Use With (Application)For use in ISO Class 5 and higher cleanroom environments, general purpose wiping and polishing of surface areas, including optics
For Use With (Equipment)For aerospace, defense electronics, food processing, computer assembly and semiconductor, circuit board, pharmaceutical and biomedical device manufacturing
Packaging Quantity300/Pk.
Unit SizePack