1000 Series Gastight™ Syringes: Luer-Tip Syringes, LT Termination
Actual product may vary
Hamilton
1000 Series Gastight™ Syringes: Luer-Tip Syringes, LT Termination
Mid-volume GASTIGHT syringe ideal for dispensing volumes from 100 μL up to 100 mL. A variety of different terminations and needle options are available in this series.
Have Questions?
Catalog NumberVolume (Metric)
FIS14-824-241 mL
Catalog number FIS14-824-24
Price (SGD)
-
Volume (Metric):
1 mL
1000 Series Syringe Terminations
Luer Tip (LT)- This termination has a PTFE, male Luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors. Also, the TLL is used with Hamilton diluters/dispensers, OEM applications, and manual operations. Autoclavable when disassembled, except on 25 mL and greater syringes. Repeated autoclaving will shorten syringe life.
Specifications
AutoclavableAutoclavable
MaterialBorosilicate Glass (Barrel), PTFE (Tip)
Diameter (English) Inner0.18 in.
Diameter (English) Outer0.355 in.
Diameter (Metric) Inner4.6 mm
Diameter (Metric) Outer9.02 mm
For Use With (Application)Ideal for manual and automated HPLC injections, commonly used in syringe pumps and other liquid handling instruments
Product TypeGastight Syringe
Max. Pressure200 psi
Max. Temperature (English)239°F
Max. Temperature (Metric)115°C
Min. Temperature (English)50°F
Model1001
Needle Point NumberNo Needle Included
QuantityEa.
Volume (Metric)1 mL
Connection TypeLuer Tip
Unit SizeEach