700 Microliter™ Syringes: LT Termination
Actual product may vary
Hamilton
700 Microliter™ Syringes: LT Termination
The 700 series syringe is the original hand-fitted Hamilton syringe. This series of syringes is ideal for dispensing volumes from 0.5 μL up to 500 μL. A variety of different terminations and needle options are available in this series.
Have Questions?
Catalog NumberVolume (Metric)Diameter (English) InnerDiameter (Metric) OuterMax. Pressure
FIS14-824-11100 μL0.061 in.7.75 mm2000 psi
Catalog number FIS14-824-11
Price (SGD)
-
Volume (Metric):
100 μL
Diameter (English) Inner:
0.061 in.
Diameter (Metric) Outer:
7.75 mm
Max. Pressure:
2000 psi
  • Original hand-fitted Hamilton syringe
  • Ideal for dispensing volumes from 0.5 µL up to 500 µL
  • A variety of different terminations and needle options are available in this series
Specifications
AutoclavableAutoclavable
MaterialPTFE (Tip), Stainless Steel (Plunger)
Diameter (English) Inner0.061 in.
Diameter (English) Outer0.305 in.
Diameter (Metric) Inner1.46 mm
Diameter (Metric) Outer7.75 mm
For Use With (Application)For dispensing low volumes to achieve the highest level of accuracy and precision possible
Product TypeMicroliter Syringe
Max. Pressure2000 psi
Max. Temperature (English)239°F
Max. Temperature (Metric)115°C
Min. Temperature (English)50°F
Model710
Needle Point NumberNo Needle Included
QuantityEa.
Volume (Metric)100 μL
Connection TypeLuer
Unit SizeEach