1700 and 1000 Series Gastight™ SampleLock™ Syringes, 22 Gauge
Actual product may vary
Hamilton
1700 and 1000 Series Gastight™ SampleLock™ Syringes, 22 Gauge
Hamilton SampleLock syringes allow you to collect, store, transport and analyze liquid or gaseous samples without the danger of evaporative loss or environmental contamination.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Max. Pressure
FIS14-815-572100 mL100 psi
FKS103100712.5 mL200 psi
Catalog number FIS14-815-572
Price (SGD)
-
Volume (Metric):
100 mL
Max. Pressure:
100 psi
Hamilton SampleLock syringes allow you to collect, store, transport and analyze liquid or gaseous samples without the danger of evaporative loss or environmental contamination.
Syringes are available in sizes ranging from 1mL to 100mL. An easy-to-use twist valve and a positive rear stop on SampleLock prevents loss of sample and plunger blowout. Optional male luer and female luer lock adapter threads on the SampleLock valve making the syringes compatible with a multitude of connectors and fittings. Typical applications are:
  • Headspace application
  • Environmental sample collection and storage
  • Pre-pressurization of gaseous sample for GC analysis
  • Injection of internal standards through the valve port into samples
  • Sample spiking
Sample Lock 1700 and 1000 Series Syringe Terminations
  • Sample Lock (SL) - The sample lock incorporates an On/Off syringe valve with RN needle. This termination is used for headspace, environmental sample collection and storage, pre-pressurization of gaseous samples for GC analysis, and sample spiking. Not autoclavable.
  • Needle Point Styles
    • Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 26-22
    Specifications
    AutoclavableNot Autoclavable
    MaterialBorosilicate Glass (Barrel), Borosilicate Glass (Barrel), PTFE (Tip), PTFE (Tip), Stainless Steel (Plunger), Stainless Steel (Plunger)
    Needle Gauge22 G
    Diameter (English) Inner1.28 in.
    Diameter (English) Inner Needle0.01 in.
    Diameter (Metric) Inner Needle0.41 mm
    Diameter (English) Outer Needle0.02 in.
    Diameter (English) Outer1.45 in.
    Diameter (Metric) Inner32.6 mm
    Diameter (Metric) Outer1.45 mm
    Diameter (Metric) Outer Needle0.72 mm
    For Use With (Application)To collect, store, transport and analyze liquid or gaseous samples without the danger of evaporative loss or environmental contamination
    Length (English) Needle2 in.
    Product TypeGeneral Purpose Syringe
    Max. Pressure100 psi
    Max. Temperature (English)176°F
    Max. Temperature (Metric)80°C
    Min. Temperature (English)50°F
    Needle Point Number2
    Min. Temperature (Metric)10°C
    QuantityEa.
    Volume (Metric)100 mL
    Product LineGastight SampleLock
    Length (Metric) Needle51 mm
    Needle Point StyleSharp, Bevel, Curved, Non-coring
    Connection TypeSample Lock, Removable
    Unit SizeEach