1000 Series Gastight™ Syringes: Luer Lock Syringes, TLL Termination
Actual product may vary
Hamilton
1000 Series Gastight™ Syringes: Luer Lock Syringes, TLL Termination
Mid-volume GASTIGHT syringe ideal for dispensing volumes from 100 μL up to 100 mL.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ModelConnection Type
FIS14-815-19425 mL1025Luer Lock
FIS14-815-19325 mL1025Luer Lock with Slots
FIS13-684-116A100 mL1100Luer Lock
FIS13-684-941 mL1001Luer Lock
FIS13-684-952.5 mL1002Luer Lock
FIS13-684-9710 mL1010Luer Lock
FIS14-815-1291 mL1001Luer Lock with Slots
FIS14-815-19550 mL1050Luer Lock
Catalog number FIS14-815-194
Price (SGD)
-
Volume (Metric):
25 mL
Model:
1025
Connection Type:
Luer Lock
Mid-volume GASTIGHT syringe ideal for dispensing volumes from 100 μL up to 100 mL.
  • PTFE Luer Lock (TLL)- This termination has a PTFE, male Luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors
  • The TLL is used with Hamilton diluters/dispensers, OEM applications, and manual operations
  • The version with slots is used during the attachment to some instruments but does not effect the connection to needles
  • Autoclavable when disassembled, except on 25mL and greater syringes; repeated autoclaving will shorten syringe life
  • Needle point styles 4 and AS are available on request)
Specifications
AutoclavableNot Autoclavable
MaterialPTFE (Tip), PTFE-tipped (Plunger)
Diameter (English) Inner0.9 in.
Diameter (English) Outer1.07 in.
Diameter (Metric) Inner23 mm
Diameter (Metric) Outer27.1 mm
For Use With (Application)Ideal for manual and automated HPLC injections, commonly used in syringe pumps and other liquid handling instruments
Product TypeGastight Syringe
Max. Pressure100 psi
Max. Temperature (English)176°F
Max. Temperature (Metric)80°C
Min. Temperature (English)50°F
Model1025
Needle Point NumberNo Needle Included
QuantityEa.
Volume (Metric)25 mL
Connection TypeLuer Lock
Unit SizeEach