1000 Series Gastight™ Syringes: Fixed-Needle Syringes, LTN Termination, 22 Gauge, 2 in. Needle Length
Actual product may vary
Hamilton
1000 Series Gastight™ Syringes: Fixed-Needle Syringes, LTN Termination, 22 Gauge, 2 in. Needle Length
Mid-volume GASTIGHT syringe ideal for dispensing volumes from 100 μL up to 100 mL. A variety of different terminations and needle options are available in this series.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Needle Point NumberModel
FIS14-815-1532.5 mL5Model 1002
FIS14-824-251 mL2Model 1001
FIS14-824-265 mL2Model: 1005
Catalog number FIS14-815-153
Price (SGD)
-
Volume (Metric):
2.5 mL
Needle Point Number:
5
Model:
Model 1002
1000 Series Syringe Terminations
  • Luer Tip Cemented Needle (LTN)- For mid volume syringes the needles are cemented into the glass syringe barrel at a point corresponding to the zero graduation mark. With this termination, dead volume is limited to the internal volume of the needle. Not autoclavable. Needle gauge is determined by the syringe volume and are not user-selectable.
    • Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 33-10
    • Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauges: 33-10
    • Point Style 5 - Conical needle with side port for penetration of septa, thin-gauged vinyls and plastics without coring. Available gauges: 26-10
Specifications
AutoclavableNot Autoclavable
MaterialBorosilicate Glass (Barrel), PTFE (Tip), Stainless Steel (Plunger)
Needle Gauge22 G
Diameter (English) Inner0.28 in.
Diameter (English) Inner Needle0.016 in.
Diameter (Metric) Inner Needle0.41 mm
Diameter (English) Outer Needle0.028 in.
Diameter (English) Outer0.4 in.
Diameter (Metric) Inner7.29 mm
Diameter (Metric) Outer10.3 mm
Diameter (Metric) Outer Needle0.72 mm
For Use With (Application)Ideal for manual and automated HPLC injections, commonly used in syringe pumps and other liquid handling instruments
Length (English) Needle2 in.
Product TypeGastight Syringe
Max. Pressure200 psi
Max. Temperature (English)122°F
Max. Temperature (Metric)50°C
Min. Temperature (English)50°F
ModelModel 1002
Needle Point Number5
QuantityEa.
Volume (Metric)2.5 mL
Needle Point StyleCone
Connection TypeLuer Tip, Cemented
Unit SizeEach