Plate Reader
Actual product may vary
Bel-Art
Plate Reader
Tool allows safe examination of the bottom of plates without risking spills on the face and eyes
Have Questions?
Catalog NumberDisplay Type
FIS14-809-175None
Catalog number FIS14-809-175
Price (SGD)
916.00
Each
Display Type:
None
Price (SGD)
916.00
Each
  • For examining cell monolayer, cell clones and for counting viral plaques or bacterial colonies
  • Easy and safe reading of hemagglutination, hemagglutination inhibition, hemolysis and agglutination tests
  • Clear deck for resting plates
  • Large, adjustable mirror is perfect for viewing plate underside
  • Mirror magnification is 3X actual plate size
  • Made of polypropylene with a glass-clear acrylic deck for placement of plates and Petri dishes
Hemagglutination, Hemagglutination Inhibition, Hemolysis and Aglutination testing
Specifications
Product TypeMicrotiter Plate Reader
Display TypeNone
Magnification3X
MaterialPolypropylene, Clear Glass Acrylic Deck
For Use With (Application)Hemagglutination, hemagglutination inhibition, hemolysis and agglutination testing
For Use With (Equipment)Microtiter plates
Unit SizeEach