1700/1000 Series Gastight™ Instrument Syringes: CX Termination
Actual product may vary
Hamilton
1700/1000 Series Gastight™ Instrument Syringes: CX Termination
Syringes are designed to fit into, or with, various different instruments such as the MICROLAB™ 500 and MICROLAB 1000
Have Questions?
Catalog NumberVolume (Metric)Diameter (Metric) InnerModel
FIS13-684-89500 μL3.26 mm1750
Catalog number FIS13-684-89
Price (SGD)
-
Volume (Metric):
500 μL
Diameter (Metric) Inner:
3.26 mm
Model:
1750
Have a PTFE plunger tip that creates a tight connection between the syringe barrel and plunger allowing for liquid or gas sampling.
  • Cheminert (CX) - This termination is a -+-28 fitting designed to fit directly into instrumentation. The X-style, meaning plunger stop, on this syringe is designed to limit wear and tear on the plunger tip.
Specifications
AutoclavableAutoclavable
MaterialBorosilicate Glass (Barrel), PTFE (Tip)
Diameter (English) Inner0.128 in.
Diameter (English) Outer0.305 in.
Diameter (Metric) Inner3.26 mm
Diameter (Metric) Outer7.75 mm
For Use With (Application)For dispensing volumes from 100μL up to 100mL
IncludesInstrument plunger stop
Product TypeGastight Syringe
Max. Pressure700 psi
Max. Temperature (English)239°F
Max. Temperature (Metric)115°C
Min. Temperature (English)50°F
Model1750
Needle Point NumberNo Needle Included
QuantityEa.
Volume (Metric)500 μL
Connection TypeChemSeal
Unit SizeEach