Personna Single-edge Prep Razor Blades
Actual product may vary
AccuTec Blades
Personna Single-edge Prep Razor Blades
Ideal for all specimen grossing applications
Have Questions?
Catalog NumberLength (Metric)
FIS12-640-187.44 cm
Catalog number FIS12-640-18
Price (SGD)
-
Length (Metric):
7.44 cm
  • Designed for the Weck handle
  • Each blade is carefully ground and honed, ensuring the sharpest possible edge
  • Cutting edges are Comfort-Coated™ for super-smooth cutting and specimen preparation
  • Blades are constructed of quality stainless steel and feature sharpness and edge retention as well as corrosion resistance
Specifications
Blade MaterialStainless Steel
Product TypePrep Razor Blade
For Use With (Application)For all Specimen Grossing Applications
Length (English)2.93 in.
Length (Metric)7.44 cm
For Use With (Equipment)Weck Handle