MeisterHand™ Mayo Dissecting Scissors
Actual product may vary
Integra Miltex
MeisterHand™ Mayo Dissecting Scissors
distal cutting
Have Questions?
Catalog NumberLength (Metric)Blade StyleMaterialProduct Type
FIS12-460-64322.9 cmStraight, BeveledStainless SteelDissecting Scissors
Catalog number FIS12-460-643
Price (SGD)
340.00
Each
Length (Metric):
22.9 cm
Blade Style:
Straight, Beveled
Material:
Stainless Steel
Product Type:
Dissecting Scissors
Price (SGD)
340.00
Each
  • Stainless steel
  • Non-glare matted finish
  • Sharp/sharp scissors available in straight and curved patterns
  • Tungsten carbide inserts seamlessly bonded to upper and lower blades and feature gold ring handles
  • SuperCut scissors have a razor sharp edge that cuts effortlessly through tissue and lower blade has micro fine serrations to help prevent tissue slippage; black ring handles
Specifications
Product TypeDissecting Scissors
Blade StyleStraight, Beveled
Length (English)9 in.
Length (Metric)22.9 cm
MaterialStainless Steel
For Use With (Application)Dissecting
Unit SizeEach