Johns Hopkins Bulldog Clamps
Actual product may vary
Integra Miltex
Johns Hopkins Bulldog Clamps
Hemostatic clamps with excellent closing pressure
Have Questions?
Catalog NumberLength (Metric)Tip Style
FIS12-460-5563.8 cmStraight
Catalog number FIS12-460-556
Price (SGD)
243.00
Each
Length (Metric):
3.8 cm
Tip Style:
Straight
Price (SGD)
243.00
Each
Available in straight and curved styles in several lengths
Specifications
Product TypeJohns Hopkins Clamp
Length (English)1.5 in.
Length (Metric)3.8 cm
MaterialStainless Steel
Tip StyleStraight
Unit SizeEach