Rochester Pean Forceps
Actual product may vary
Rochester Pean Forceps
Rochester Pean Forceps
Integra Miltex
Rochester Pean Forceps
Straight
Have Questions?
Catalog NumberLength (Metric)
FIS12-460-548305 mm
Catalog number FIS12-460-548
Price (SGD)
611.00
Each
Length (Metric):
305 mm
Price (SGD)
611.00
Each
  • Full horizontal serrations
Specifications
Tip StyleStraight
Length (English)12 in.
Length (Metric)305 mm
Product TypeForceps, Rochester Pean
Quantity (No. per Case)Each
IncludesHorizontal serrations
Unit SizeEach