▸
Facility Safety and Maintenance
▸
Hand Tools and Power Tools
▸
Scissors
▸
Micro Iris Scissors
Actual product may vary
Integra Miltex
Micro Iris Scissors
For fine cutting
Have Questions?
Catalog Number
Blade Style
FIS12-460-280
Straight
Catalog number
FIS12-460-280
Price (SGD)
1,437.00
Each
Add to cart
Blade Style:
Straight
Price (SGD)
1,437.00
Each
Add to cart
High-quality surgical-grade stainless steel construction provides optimal performance
Fully tempered precision screw locks maintain perfect alignment
Ergonomic design combines weight, balance and feel for optimal comfort and performance
Well rounded and polished ring handles provide exceptional comfort and reduced hand fatigue
Sharp points
Length: 4 in. (10.2cm)
Specifications
Product Type
Micro Iris Scissors
Blade Style
Straight
Grade
Premium
Length (English)
4 in.
Length (Metric)
10.16 cm
Material
Stainless Steel
For Use With (Application)
Surgical, ophthalmic
Tip Style
Sharp/Sharp