Magic Touch 2™ Ice Buckets
Actual product may vary
Bel-Art
Magic Touch 2™ Ice Buckets
Superior insulating properties combined with high chemical resistance
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)ColorHeight (English)
FIS11-999-0562.5 LRed7.06 in.
FIS11-999-0552.5 LBlack7.06 in.
FIS11-999-0614 LGreen9 in.
Catalog number FIS11-999-056
Price (SGD)
377.00
Each
Capacity (Metric):
2.5 L
Color:
Red
Height (English):
7.06 in.
Price (SGD)
377.00
Each
Superior insulating properties combined with high chemical resistance
  • Compatible not only with wet ice, but also with ultra-low temperature materials including dry ice, salt slurries, dry ice solvent slurries or liquid nitrogen
  • Able to withstand a wide temperature range of -196°C (-320°F) to 100°C (212°F)
  • High-quality, expanded urethane laboratory ice buckets are lightweight, durable, nonsweating and impervious to moisture and odors
  • Convenient pour spout, drip-proof rims, a fill line indicator
  • Snug-fitting keyed lid and base for easy stacking
  • Lightly textured, nonslip finish and raised edge provides secure transport and a stylish look
Specifications
Height (Metric)17.9 cm
Height (English)7.06 in.
Diameter (Metric)26.8 cm
Capacity (English)0.56 gal.
IncludesPour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base
DesignLightly textured, Non-slip
MaterialUrethane
Diameter (English)10.56 in.
TypeIce Bucket
ColorRed
Capacity (Metric)2.5 L
Holds24 Microtubes
AutoclavableNot Autoclavable
Temperature ControlNo
Temperature Range-196°C to +100°C
Unit SizeEach