2-Hydroxy-5-iodopyridine, 97%
2-Hydroxy-5-iodopyridine, 97%
2-Hydroxy-5-iodopyridine, 97%
Bel-Art
2-Hydroxy-5-iodopyridine, 97%
CAS: 13472-79-2 | C5H4INO | 220.997 g/mol
Have Questions?
Catalog NumberQuantity
FIS11-394-4565 g
Catalog number FIS11-394-456
Price (SGD)
141.00
Each
Quantity:
5 g
Request bulk or custom format
Price (SGD)
141.00
Each
Specifications
Chemical Name or Material2-Hydroxy-5-iodopyridine
CAS13472-79-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Employed as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Employed as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Light Sensitive. Store in dark. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only