Exhaust Fume Hood
Actual product may vary
Bel-Art
Exhaust Fume Hood
Exhaust fume hood removes fumes and vapours to provide secure work environment for venting tanks and vats.
Have Questions?
Catalog NumberFilter Type
FIS11-391-121Unfiltered
Catalog number FIS11-391-121
Price (SGD)
3,141.00
Each
Filter Type:
Unfiltered
Price (SGD)
3,141.00
Each
  • Rests on rear edge on tank to draw air across the top away from operator
  • Tapered
  • Made from non-corrosive polypropylene
  • Available in different sizes
H50015-0000:
  • Width x Height: 24 x 12 in. (61 x 31cm)
  • Slot Height x Depth: 3 x 3 in. (76 x 76cm
  • Stack O.D.: 6 in. (15mm)
  • Material thickness: 0.187 in. (4.8mm)
H50016-0000:
  • Width x Height: 36 x 18 in. (91 x 46cm)
  • Slot Height x Depth: 3 x 3 in. (76 x 76cm
  • Stack O.D.: 8 in. (20mm)
  • Material thickness: 0.187 in. (4.8mm)
H50016-0000:
  • Width x Height: 48 x 24 in. (122 x 61cm)
  • Slot Height x Depth: 3 x 4 in. (76 x 102cm)
  • Stack O.D.: 8 in. (20mm)
  • Material thickness: 6.4 in. (0.25mm)
Dying, chemical processing, blacktop and hot mix processing, and leather tanning
Specifications
Product TypeFume Hood
CoatingUncoated
Filter TypeUnfiltered
Lighting TypeNone
MaterialPolypropylene
Depth (English)3 in.
Depth (Metric)7.6 cm
Dimensions (L x W x H)61 x 31 x 7.6 cm (24 x 12 x 3 in.)
Height (English)12 in.
Height (Metric)31 cm
LocationLaboratory, Classroom
Width (English)24 in.
Width (Metric)61 cm
Unit SizeEach