Chemware™ PTFE Boiling Stones
Actual product may vary
Saint-Gobain
Chemware™ PTFE Boiling Stones
Use these PTFE chips to promote gentle, efficient boiling and help prevent overheating of highly volatile liquids. Chemware™ PTFE Boiling Stones are not affected by acids, hydrocarbons, ketones, esters, alcohols or alkalis.
Have Questions?
Catalog NumberQuantity
FIS09-191-2016 oz. (450 g)
Catalog number FIS09-191-20
Price (SGD)
-
Quantity:
16 oz. (450 g)
Use these PTFE chips to promote gentle, efficient boiling and help prevent overheating of highly volatile liquids. Saint Gobain Performance Plastics™ Chemware™ PTFE Boiling Stones are not affected by acids, hydrocarbons, ketones, esters, alcohols or alkalis.
  • Use these PTFE chips to promote gentle, efficient boiling and help prevent overheating of highly volatile liquids.
  • Saint Gobain Life Sciences Chemware™ PTFE Boiling Stones are not affected by acids, hydrocarbons, ketones, esters, alcohols or alkalis.
  • Plastic Bottle
Specifications
TypeBoiling Stone
MaterialPTFE
DescriptionBoiling Stones
For Use With (Equipment)Distillation Columns
Quantity16 oz. (450 g)
Unit SizeEach