Dry-Keeper™ Desiccator Cabinet
Actual product may vary
Bel-Art
Dry-Keeper™ Desiccator Cabinet
Clear, durable polystyrene desiccator cabinets have black ABS accents and movable shelves for adjusting dry storage space configuration for moisture-sensitive materials.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorCabinet StyleNo. of ShelvesHolds
FIS08-647-24TranslucentVertical3 Adjustable Perforated70 Cans
FIS08-647-26ClearVertical2 Adjustable70 Cans
FIS08-647-28TranslucentHorizontal2 Adjustable Perforated78 Cans
Catalog number FIS08-647-24
Price (SGD)
-
Color:
Translucent
Cabinet Style:
Vertical
No. of Shelves:
3 Adjustable Perforated
Holds:
70 Cans
  • Generous 2 cu. ft. interior volume
  • Adjustable, removable, and perforated shelves facilitate full circulation of air; additional shelves and rails sold separately
  • Large, in-door dial hygrometer for easy visualization of RH%
  • Gasketed door closes with two latches for a tight seal
  • Standard cabinets include desiccant tray (desiccant sold separately)
Horizontal Cabinets
  • Include two shelves
  • Shelf rails can be repositioned on any one of eight steps set at 1.1 in. (28 mm) intervals
  • Interior opening measures 17.75 x 9 in. (45 x 23 cm)
Vertical Cabinets
  • Includes three shelves
  • Shelf rails can be repositioned on any one of 15 steps set at 1.1 in. (28 mm) intervals
  • Interior opening measures 10 x 17 in. (25 x 43 cm)
Autodesiccator (H42058-0003, H42056-1003)
  • 120VAC electronic dehumidification system maintains relative humidity levels at 30 to 40%†; keeps dry air circulating throughout the cabinet
  • Permanent desiccant contained in the control unit
  • Red light indicates operation of the 4.5 hour absorption period; green light indicates operation of the 0.5 hour regeneration of the desiccant
Specifications
ColorTranslucent
Cabinet StyleVertical
MaterialMethyl Methacrylate
Product TypeDesiccator
Door StyleHinged and gasketed
For Use With (Application)Adjusting dry storage space configuration for moisture-sensitive materials, can be used with Scienceware™ desiccant cartridges, canisters or similar desiccant
Holds70 Cans
IncludesBlack ABS accents, 3 adjustable perforated shelves and shelf rails, desiccant tray and dial hygrometer, 2 metal hinges and latches, door-mounted hygrometer
No. of Shelves3 Adjustable Perforated
Volume (English)2 cu. ft.
Length (English)12.5 in.
Length (Metric)31.75 cm
Width (English) Door9.8 in.
Width (Metric) Door25 cm
Height (English)20 in.
Height (English) Opening17.7 in.
Height (Metric)51 cm
Depth (English)15.5 in.
Depth (Metric)39.3 cm
Unit SizeEach