Mini Magnifier
Actual product may vary
Bel-Art
Mini Magnifier
Provides hands-free 1.75X magnification of slides, plates or gels placed on the Mini Light Box II
Have Questions?
Catalog NumberMagnification
FIS07-910-131.75x
Catalog number FIS07-910-13
Price (SGD)
490.00
Each
Magnification:
1.75x
Price (SGD)
490.00
Each
  • Tilts for optimal viewing
  • Measures 10.5 x 14.3 x 17.2cm H
  • Also useful for viewing of x-ray films, stained gels, slides or text
Specifications
For Use With (Application)Magnified observation of petri dishes, slides, gels or text
For Use With (Equipment)Scienceware™ Mini Light Box II
Height (English)6.7 in.
Height (Metric)17.2 cm
Length (English)6.25 in.
Length (Metric)15.8 cm
Weight (English)13 oz.
Weight (Metric)368.5 g
Width (English)5.9 in.
Width (Metric)14.9 cm
Diameter (Englis) Lens5.5 in.
Diameter (Metric) Lens140 mm
Magnification1.75x
MaterialGlass Lens
Product TypeMagnifier, Mini
Unit SizeEach