▸
Filters and Filtration
▸
Capsules and Inline Filters
▸
Precolumn Inline Filters
Actual product may vary
Idex
Precolumn Inline Filters
Protect larger columns and injections from harmful particulates
Have Questions?
Catalog Number
Pore Size
Housing Material
Volume (Metric)
FIS05-707-31
0.5 μm
Stainless Steel
0.66 uL
Catalog number
FIS05-707-31
Price (SGD)
-
Pore Size:
0.5 μm
Housing Material:
Stainless Steel
Volume (Metric):
0.66 uL
Ideal for higher-flow systems
Interchangeable frits make protection more economical
Compatible with SFC and HPLC
Available to fit a variety of tubing sizes
Specifications
Pore Size
0.5 μm
Housing Material
Stainless Steel
Type
Precolumn Inline Filter
Volume (Metric)
0.66 uL
Length (English)
0.94 in.