▸
Chromatography Systems
▸
Liquid Chromatography Instruments
▸
Individual HPLC Components and Accessories
▸
HPLC Solvent Handling Component Accessories
▸
Inlet Solvent Filters
Actual product may vary
Idex
Inlet Solvent Filters
Stainless-steel or multiple materials
Have Questions?
Catalog Number
Material
FIS05-707-24
Stainless Steel
Catalog number
FIS05-707-24
Price (SGD)
-
Material:
Stainless Steel
Cups and end fittings sold separately, see 05-704-54 and 045-707-83.
Specifications
Flow Rate
40 mL/min.
Includes
1 Filter (Cups and end fittings sold separately)
Material
Stainless Steel
Pore Size
10 μm
Thread Size
N/A
Type
Inlet Solvent Filter