PEEK Chromatography Tubing
Actual product may vary
Idex
PEEK Chromatography Tubing
An excellent replacement for stainless steel tubing
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English) InnerOverall Length (English)Maximum PressureColor
FIS05-701-190.005 in.100 ft.7000 psiNatural
FKS130298920.03 in.5 ft.7000 psi (483 bar)Green
FIS05-701-120.01 in.50 ft.7000 psi (483 bar)Blue
Catalog number FIS05-701-19
Price (SGD)
-
Diameter (English) Inner:
0.005 in.
Overall Length (English):
100 ft.
Maximum Pressure:
7000 psi
Color:
Natural
An excellent replacement for stainless steel tubing
Can be used in place of stainless steel tubing in most liquid analytical systems
  • Biocompatible, inert and easy to cut
  • Great for high-pressure applications
  • Wide choice of outer and inner diameters
  • Many sizes color-coded for easier inner diameter identification
Specifications
TypeTubing
ColorNatural
For Use With (Application)High-Pressure Applications
Diameter (English) Inner0.005 in.
Overall Length (English)100 ft.
Maximum Pressure7000 psi
Unit SizeEach