Flangeless Fittings: Ferrules, Tefzel
Actual product may vary
Idex
Flangeless Fittings: Ferrules, Tefzel
No need to flange tubing
Have Questions?
Catalog NumberColorFor Use With (Equipment)Length (English)Width (English)
FIS05-700-77Natural1/16 in. OD Tubing, Male Nuts0.22 in.0.18 in.
Catalog number FIS05-700-77
Price (SGD)
-
Color:
Natural
For Use With (Equipment):
1/16 in. OD Tubing, Male Nuts
Length (English):
0.22 in.
Width (English):
0.18 in.
No need to flange tubing
Excellent for connecting rigid or semi-rigid tubing
Specifications
TypeFlangeless Ferrule
Length (English)0.22 in.
Width (English)0.18 in.
MaterialETFE
ColorNatural
For Use With (Equipment)1/16 in. OD Tubing, Male Nuts
Unit SizePack