Nalgene™ Graduated HDPE Bucket
Nalgene™ Graduated HDPE Bucket
Actual product may vary
Nalgene™ Graduated HDPE Bucket
Nalgene™ Graduated HDPE Bucket
Nalgene™ Graduated HDPE Bucket
Thermo Scientific
Nalgene™ Graduated HDPE Bucket
Transfer laboratory solutions and powders with the Thermo Scientific™ Nalgene™ Graduated HDPE Bucket, featuring a wide-lipped pour spout and polyethylene-coated handle.
Have Questions?
Catalog NumberCapacity (Metric)
FIS03-687-309.5 L
Catalog number FIS03-687-30
Price (SGD)
337.00
EA
Add to cart
Capacity (Metric):
9.5 L
Price (SGD)
337.00
EA
Add to cart
Transfer laboratory solutions and powders with the Thermo Scientific™ Nalgene™ Graduated HDPE Bucket, featuring a wide-lipped pour spout and polyethylene-coated handle. This heavy-duty, rugged high density polyethylene bucket is chemically compatible with a wide range of lab reagents.
  • High-density polyethylene construction offers more durability than hard rubber
  • Polyethylene-coated wire bail handle provides a comfortable and sturdy grip
  • Molded-in graduations in pints and liters are easy to read
Recycling information:
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in. 
Specifications
Capacity (Metric)9.5 L
GraduationsMolded
MaterialHDPE
Capacity (English)2.5 gal.
Description2.5 gal. (9.5 L) Bucket
Handle MaterialPolyethylene Coated
IncludesHandle
Quantity1 Ea.
ShapeWide-Lipped Pour Spout
Unit SizeEA