Wheaton™ M-T Vial File™ and Accessories
Wheaton™ M-T Vial File™ and Accessories
Actual product may vary
DWK Life Sciences
Wheaton™ M-T Vial File™ and Accessories
For compact and organized sample storage
Have Questions?
Catalog NumberProduct TypeQuantityFor Use With (Equipment)
FIS02-912-821Storage Box6 Case24 x 20mL vials
Catalog number FIS02-912-821
Price (SGD)
550.00
CS
Add to cart
Product Type:
Storage Box
Quantity:
6 Case
For Use With (Equipment):
24 x 20mL vials
Price (SGD)
550.00
CS
Add to cart
For compact and organized sample storage
  • Stores vials in clear polystyrene case
  • Foam inserts and partitions provide protection
  • With alphanumerical indexing card for identification
  • Index cards and partitions also available separately
Specifications
Closure TypeHinged Lid
ColorClear
MaterialPlastic
ShapeRectangular
For Use With (Application)Convenient Storage and Transportation of Sample Vials
For Use With (Equipment)24 x 20mL vials
Holds24 (2, 4, 20 mL Vials)
IncludesFoam Insert, Index Card, Partition, Polystyrene Case
No. of Places24
Product TypeStorage Box
Quantity6 Case
Unit SizeCS