Fisherbrand™ Low- and Tall-Form PTFE Evaporating Dishes
Fisherbrand™ Low- and Tall-Form PTFE Evaporating Dishes
Actual product may vary
Fisherbrand™ Low- and Tall-Form PTFE Evaporating Dishes
Inert and usable to 280°C (536°F).
Have Questions?
Catalog NumberCapacity (Metric)Diameter (Metric)
FIS02-617-14050 mL40 mm
Catalog number FIS02-617-140
Price (SGD)
99.00
EA
Add to cart
Capacity (Metric):
50 mL
Diameter (Metric):
40 mm
Price (SGD)
99.00
EA
Add to cart
  • Inert and usable to 280°C (536°F).
  • Smooth internal finish.
  • With heavy walls
  • With pouring spout
Specifications
DescriptionEvaporating Dish Tall PTFE 50 mL
Capacity (Metric)50 mL
MaterialPTFE
ShapeRound
TypeEvaporating Dish
For Use With (Application)Evaporating Application
Interior FinishSmooth
Temperature Range (English)500°F
Temperature Range (Metric)260°C
Length (English)2 in.
Length (Metric)51.84 mm
Height (Metric)50 mm
Height (English)1.96 in.
Diameter (English)1.57 in.
Diameter (Metric)40 mm
Unit SizeEA