PYREX™ Tall-Form Berzelius Beakers with Spout, Graduated
PYREX™ Tall-Form Berzelius Beakers with Spout, Graduated
Actual product may vary
PYREX™ Tall-Form Berzelius Beakers with Spout, Graduated
PYREX™ Tall-Form Berzelius Beakers with Spout, Graduated
PYREX™ Tall-Form Berzelius Beakers with Spout, Graduated
Graduated to indicate approximate content
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS02-546G1 L
FIS02-546B200 mL
FIS02-546C300 mL
FIS02-546E500 mL
Catalog number FIS02-546G
Price (SGD)
287.00
PK
Add to cart
Capacity (Metric):
1 L
Price (SGD)
287.00
PK
Add to cart
Corning™ PYREX™ Tall-Form Berzelius Beakers are graduated to indicate approximate content.
A tall beaker with spout to facilitate pouring. For convenience, these beakers are graduated to indicate their approximate content.
Specifications
Capacity (English)33.81 oz.
Diameter (English) Outer1.53 in.
Diameter (Metric) Outer8.9 cm
Capacity (Metric)1 L
MaterialBorosilicate Glass
For Use With (Application)Convenient for Easy Pouring
Graduation Range50 to 950 mL
Height (English)7.36 in.
Height (Metric)18.7 cm
IncludesSpout
Increments50 mL
Label TypeMarking Spot
Quantity6 Pk.
TypeBeaker
DisposableReusable
GraduatedYes
Certifications/ComplianceASTM E960
ScaleWhite Enamel Scale
StyleBerzelius
Unit SizePK