Tyvek™ 400 Hooded Coveralls with Skid-Resistant Boots
Actual product may vary
Tyvek™ 400 Hooded Coveralls with Skid-Resistant Boots
Tyvek™ 400 Hooded Coveralls with Skid-Resistant Boots
Tyvek™ 400 Hooded Coveralls with Skid-Resistant Boots
Tyvek™ 400 Hooded Coveralls with Skid-Resistant Boots
Tyvek™ 400 Hooded Coveralls with Skid-Resistant Boots
DuPont
Tyvek™ 400 Hooded Coveralls with Skid-Resistant Boots
Coveralls with attached skid-resistant boots include stormflaps and a respirator fit hood.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSizeSize (English) ChestWidth (English) ChestHeight (English)
FIS01-361-2CLarge39.75 to 43.25 in.26.5 in.5.5 to 5.9 ft.
FIS01-361-2BMedium36.75 to 40.25 in.25 in.5.3 to 5.7 ft.
FIS01-361-2DX-Large42.75 to 46.25 in.28 in.5.8 to 6.2 ft.
Catalog number FIS01-361-2C
Price (SGD)
722.00
Case
Size:
Large
Size (English) Chest:
39.75 to 43.25 in.
Width (English) Chest:
26.5 in.
Height (English):
5.5 to 5.9 ft.
Price (SGD)
722.00
Case
Comfort fit design based on extensive wearer input to provide a comfortable garment design that enables a greater range of movement while stretching and bending.
  • Comfort fit design provides a more tailored fit, offers reinforcement in high stress areas for fewer blowouts
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Attached Tyvek FC boots have special coating to provide added skid-resistance
  • Longer zipper extends to chin for complete coverage of neck area and provides easeir donning and doffing
  • Elastic opening for tighter fit at wrist
  • Elastic waist to better position garment
  • Elastic ankles
  • Serged seam
  • White
Specifications
Product TypeGeneral Purpose Coveralls
SizeLarge
GenderUnisex
MaterialTyvek
ColorWhite
DisposableLimited-use
Waist StyleElastic
Ankle StyleElastic, Attached Boots
Cuff StyleElastic
Neck StyleHooded
Hood TypeElastic, Respirator Fit
Closure TypeFront Zipper
Seam TypeSerged
SterilityNon-sterile
ResistanceParticle
ModelTY122SWH
Size (English) Chest39.75 to 43.25 in.
Size (English) Inseam29 in.
Size (Metric) Chest100.9 to 109.8 cm
Size (Metric) Inseam73.6 cm
Thickness5.9 mil
For Use With (Application)Lead and Asbestos Abatement, Remediation, General Maintenance and Operations, Spray Painting, General Cleaning
Length (English) Sleeve35 in.
Length (Metric) Sleeve88.9 cm
Width (English) Chest26.5 in.
Width (Metric) Chest67.3 cm
Height (English)5.5 to 5.9 ft.
Height (Metric)1.6 to 1.8 m
Quantity25/Cs.
Unit SizeCase