Traceable™ Alarm RH/Temperature Monitor
Traceable™ Alarm RH/Temperature Monitor
Actual product may vary
Traceable™ Alarm RH/Temperature Monitor
Monitors round-the-clock for critical experimental requirements.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
CTL15-079-630Internal temp/humidity/sensors
CTL14-649-84External temp/humidity sensors
Catalog number CTL15-079-630
Price (SGD)
206.00
EA
Add to cart
Includes:
Internal temp/humidity/sensors
Price (SGD)
206.00
EA
Add to cart
Triple-display and remote control unit offers safety and convenience. Fisherbrand™ Traceable™ Remote Alarm RH/Temperature Monitors are ideal for routine measurements, round-the-clock monitoring, quality control and critical experiments. Place sensor in hoods, cleanrooms, coolers, stockrooms, incubators, environmental chambers, and chemical storage areas.
  • Multi-point calibration on an individually-numbered Traceable™ certificate
  • Triple display simultaneously shows minimum/maximum and current humidity
  • Minimum and maximum memory feature for both temperature and humidity
  • Internal or external ambient sensor reads temperature and humidity
  • Alarm sounds when reading rises above or falls below set points
  • Supplied with flip-open stand for lab bench and slot for wall mounting
  • High-impact, chemical-resistant ABS plastic construction
Specifications
TypeRemote Alarm Rh/Temperature Monitor
Humidity Range5 to 95%
ResolutionHumidity Res: 0.1°
AccuracyHumidity Accuracy: ±3% midrange, ±5% otherwise
Temperature Range (Metric)0°C to 60°C
Temperature Accuracy±0.5°C
Alarm TypesAudible
Display TypeLCD w/ 0.5 in. digits
MaterialABS Plastic
Temperature Range (English)32°F to 140°F
Temperature Resolution0.1°F
Dimensions (L x W x H)2.75 x 4.25 x 0.75 in. (7 x 10.8 x 1.9 cm)
Length (English)2.75 in.
Length (Metric)7 cm
Width (English)4.25 in.
Width (Metric)10.8 cm
Height (English)0.75 in.
Height (Metric)1.9 cm
Weight (English)4 oz.
Weight (Metric)0.113 kg
For Use With (Application)For routine measurements, round-the-clock monitoring, quality control and critical experiments, place sensor in hoods, cleanrooms, coolers, stockrooms, incubators, environmental chambers and chemical storage areas
IncludesInternal temp/humidity/sensors
Certifications/ComplianceISO 17025, A2LA, NIST
Unit SizeEA