Traceable™ Refrigerator/Freezer Alarm Thermometers
Traceable™ Refrigerator/Freezer Alarm Thermometers
Actual product may vary
Traceable™ Refrigerator/Freezer Alarm Thermometers
Provides accurate reading when freezer/refrigerator doors are open with temperature-buffered, sensor-sealed in bottled glycol solution
Have Questions?
Catalog NumberTemperature Range (Metric)
CTL06-664-23-50°C to +70°C
Catalog number CTL06-664-23
Price (SGD)
115.00
Special offer
Ends: 31-Dec-2025
198.00
Save 83.00 (42%)
EA
Add to cart
Temperature Range (Metric):
-50°C to +70°C
Price (SGD)
115.00
Special offer
Ends: 31-Dec-2025
198.00
Save 83.00 (42%)
EA
Add to cart
Provides accurate reading when freezer/refrigerator doors are open with temperature-buffered, sensor-sealed in bottled glycol solution. Fisher Scientific™ Traceable™ Refrigerator/Freezer Alarm Thermometer shows current, high and low temperatures and alerts user with visual and audible signals if temperature diverts from set points.
  • Monitors temperature in freezers, refrigerators, incubators, water baths and heating blocks
  • Triple display simultaneously shows current, high and low temperatures
  • Alarm (Cat. No. 06-664-11 only) set in 1° increments alerts user with visual and audible signals if temperature goes outside setpoints
  • 10 ft. long (3m) micro cable permits doors to close on it
  • Dimensions: 4.25L x 2.75W x 0.75 in.H (11 x 7 x 2cm)
Specifications
Certifications/ComplianceISO 17025, A2LA, NIST
Dimensions (L x W x H)2.5 x 3.25 x 0.63 in. (63.5 x 82.5 x 16 mm)
For Use With (Application)Monitoring temperature in freezers, refrigerators, incubators, heating blocks
Height (English)0.63 in.
Height (Metric)16 mm
IncludesVelcro™ and magnetic mounting strip, one AAA battery, ABS Plastic case
Length (English) Overall2.5 in.
Length (Metric)63.5 mm
Length (Metric) Overall63.5 mm
MaterialPlastic
Resolution0.1°
Weight (English)4 oz.
Weight (Metric)0.11 kg
Width (English)3.25 in.
Product TypeRefrigerator/Freezer Thermometer
DescriptionBottle Probe
Max. Temperature (English)158°F
Max. Temperature (Metric)70°C
Min. Temperature (English)-58°F
Min. Temperature (Metric)-50°C
Temperature Range (English)-58°F to +158°F
Temperature Range (Metric)-50°C to +70°C
CalibratedYes
For Use With (Equipment)Monitoring Temperature in Freezer, Refrigerator, Incubator, Heating Block
Accuracy±0.5°C
DisplayDigital LCD
Width (Metric)82.5 mm
Unit SizeEA